Learn More
Carolina™ Protozoa Survey Set

Shop All Carolina Biological Supply Products


Includes
- Amoeba
- Euglena
- Paramecium
- Stentor
- Volvox
- Spirostomum
- Instructions
- Six individually labeled cultures
- Each culture can be used for a class of 30
Specifications
For Use With (Application)
Study protists, cell structure and diversity.
Storage Requirements
20°C to 22°C
Quantity
Ea.
Includes
Amoeba, Euglena, Paramecium, Stentor, Volvox, Spirostomum, instructions
Sufficient For
Class of 30 students
Product Type
Protozoa Survey Set
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.