Learn More
World Precision Instrument Cartridge For WPSS01
SDP
Supplier:  World Precision Instrument WPSS02
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
The WPSS01 skin stapler is an affordable, reliable solution for veterinary wound closures. A skin stapler is designed for the efficient closure of surgical incisions or wounds by applying surgical staples to the skin, reducing surgery time and time under anesthesia. This handheld instrument features reloadable cassettes which are sold separately. The clear handle, which is autoclavable, allows you to spot contamination. The angled tip design gives you a clear view of the incision line. Pre-loaded, sterile staple cassettes containing 35 staples made loading efficient, and minimizes the chance of infection at the wound site. The lightweight and durable design is also ergonomic, making it easy to load and simple to use. The WPI skin stapler is made in the USA.
Catalog No. 50-253-8253
missing translation for 'QuantityLeft'