Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carvedilol, Tocris Bioscience™

Catalog No. 268550 Shop All R&D Systems Products
Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and non-selective β antagonist; also potent α1 antagonist

Potent β-adrenoceptor and α1-adrenoceptor antagonist (Ki values are 0.81, 0.96 and 2.2 nM for β1-, β2- and α1-adrenoceptors respectively) that displays antihypertensive and peripheral vasodilatory activity. Blocks cardiac inward-rectifier K+ (KIR) channels, voltage-dependent Ca2+ channels and exhibits antioxidant properties at higher concentrations.

Chemical Identifiers

CAS 72956-09-3
Molecular Formula C24H26N2O4
Molecular Weight (g/mol) 406.48
MDL Number MFCD00864692,MFCD00869663
InChI Key OGHNVEJMJSYVRP-UHFFFAOYNA-N
Synonym carvedilol, coreg, dilatrend, carvedilolum, eucardic, carvedilolum latin, kredex, querto, 1-9h-carbazol-4-yloxy-3-2-2-methoxyphenoxy ethyl amino-2-propanol, 1-9h-carbazol-4-yl oxy-3-2-2-methoxyphenoxy ethyl amino propan-2-ol
PubChem CID 2585
ChEBI CHEBI:3441
IUPAC Name 1-(9H-carbazol-4-yloxy)-3-{[2-(2-methoxyphenoxy)ethyl]amino}propan-2-ol
SMILES COC1=CC=CC=C1OCCNCC(O)COC1=CC=CC2=C1C1=C(N2)C=CC=C1

Specifications

Quantity 50 mg
Formula Weight 406.47
Percent Purity 98%
Chemical Name or Material Carvedilol
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.