Learn More
3M™ Cashew Protein Rapid Kit L25CHW, Sold by AquaPhoenix Scientific
Shop All AquaPhoenix Scientific Products

- Results visible at 11 ± 1 min. after application of the sample
- Rapid, qualitative immunochromatographic lateral flow test cartridges designed for the detection of allergenic food proteins
- Designed to detect both processed and unprocessed allergen proteins
- Test can accommodate environmental swab, CIP rinse water and food samples
Specifications
Format
Kit
Product Type
Microbiology Media
Test Type
Allergen
For Use With (Application)
Screening cashew proteins in CIP final rinse water, environmental swab samples, food ingredients and processed food products
Quantity
25/Cs.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.