Learn More
Enzo Life Sciences Catalase fluorometric detection kit (500tests)
SDP
Supplier:  Enzo Life Sciences ADI907027
Encompass_Preferred
The Catalase fluorometric detection kit provides a higher throughput sensitive method for measuring catalase activity in just one hour. This is a homogeneous mix-and-read assay that enables you to monitor multiple time points for a kinetic read. Positive control is provided. Sensitivity: (range 0- 4 U/ml). UniProt ID: P00432. Applications: Fluorescent detection, HTS . Species reactivity: Species independent . Content: Reaction buffer concentrate, Detection Reagent, Horseradish peroxidase concentrate, Hydrogen peroxide, Catalase enzyme.
Catalog No. 50-200-7085
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.