Learn More
Cathepsin K Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$158.00 - $487.50
Specifications
Antigen | Cathepsin K |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17987820
![]() |
Novus Biologicals
NBP17987820UL |
20 μL |
Each for $158.00
|
|
|||||
NBP179878
![]() |
Novus Biologicals
NBP179878 |
100 μL |
Each for $487.50
|
|
|||||
Description
Cathepsin K Polyclonal specifically detects Cathepsin K in Human samples. It is validated for Western Blot.Specifications
Cathepsin K | |
Polyclonal | |
Rabbit | |
Cancer | |
NP_000387 | |
1513 | |
Synthetic peptide directed towards the middle region of human CTSK. Peptide sequence SPQNLVDCVSENDGCGGGYMTNAFQYVQKNRGIDSEDAYPYVGQEESCMY. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
RUO | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
cathepsin K, cathepsin K (pycnodysostosis), Cathepsin O, cathepsin O1, Cathepsin O2, Cathepsin X, CTS02, CTSO, CTSO1, CTSO2, EC 3.4.22, EC 3.4.22.38, MGC23107, PKND, PYCD | |
CTSK | |
IgG | |
Affinity Purified | |
37 kDa |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.