Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CBS Scientific Horizontal Productivity Packages

Allows casting second gel while first is running for increased efficiency.
Supplier: CBS Scientific GCHSU020
Description
CBS Economy Horizontal Productivity Packages feature platinum electrode cassettes which provide rapid sample velocity and clear focused bands without smiling and cab be easily removed for cleaning or replacement.Gel Tray Specifications:
- Winged gel trays for easy handling - no more sticking fingers into slippery buffer to extract gel tray
- Made from sturdy 6.4mm thick UV transparent acrylic
- Grooves in gel tray corners create agarose anchors to prevent gel flotation during electrophoresis
- Gel tray configuration allows single run or multiple simultaneous short runs
- Three options for gel casting; tapered baffles, External Gel Casters and Continuously Adjustable Caster
- Adjustable height polycarbonate or PTFE-coated aluminum combs are available in many configurations, with or without markers
- PTFE-coated combs allow faster well formation and no wells are lost when using low percentage or low melt agarose
- Buffer recirculation ports allow maintenance of uniform pH and ionic strength and reduce frequency of buffer changes necessary during long runs
- Tight-fitting safety cover reduces condensation during electrophoresis
- Coolant can be recirculated beneath gel bed to provide constant temperature runs
- Combs are printed with catalog numbers and thickness for easy identification
Specifications
CE | |
Compatible Gel Tray: WGT-020 | |
43 x 21 x 10 cm (16.9 x 8.26 x 3.9 in.) | |
L x W: 20 x 20 cm (7.8 x 7.8 in.) | |
3 Years |
Blue | |
Gel Tray I.D.: 18.7 x 20 cm (7.3 x 7.8 in.) | |
Gel Electrophoresis System | |
1850 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction