Learn More
ATCC CCD-112CoN; Colon Fibroblast; Human (Homo sapiens)
Supplier:  ATCC CRL1541
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
CCD-112CoN is a cell line exhibiting fibroblast morphology that was isolated from the large intestine of a 22-week-old, female patient. This cell line was deposited by ATCC.
Catalog No. 50-238-2136
missing translation for 'QuantityLeft'