Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCT 018159, Tocris Bioscience™

Catalog No. p-4783835 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Hsp90 inhibitor

Novel inhibitor of heat shock protein 90 (Hsp90) ATPase activity (IC50 = 5.7 μM) that displays selectivity over human Hsp72 and topoisomerase II. Inhibits proliferation of HCT116 human colon tumor cells and produces upregulation of Hsp70 and downregulation of c-Raf and cdk4. More soluble than 17-AAG (Cat. No. 1515) and is independent of NQO1/DT-diaphorase and P-glycoprotein expression.

Chemical Identifiers

CAS 171009-07-7
Molecular Formula C20H20N2O4
Molecular Weight (g/mol) 352.39
InChI Key BBTMGPQSLSYPHE-UHFFFAOYSA-N
Synonym 6-4-2,3-dihydro-1,4-benzodioxin-6-yl-5-methyl-1,2-dihydropyrazol-3-ylidene-4-ethyl-3-hydroxycyclohexa-2,4-dien-1-one, kinome_3591, owpmenvyxdjdow-uhfffaoysa-n, 4-4-2,3-dihydro-benzo 1,4 dioxin-6-yl-5-methyl-1h-pyrazol-3-yl-6-ethyl-benzene-1,3-diol
PubChem CID 6749166
IUPAC Name 6-[4-(2,3-dihydro-1,4-benzodioxin-6-yl)-5-methyl-1,2-dihydropyrazol-3-ylidene]-4-ethyl-3-hydroxycyclohexa-2,4-dien-1-one
SMILES CCC1=CC(=C2C(=C(NN2)C)C3=CC4=C(C=C3)OCCO4)C(=O)C=C1O

Specifications

Quantity 10 mg
Formula Weight 352.39
Percent Purity >98%
Chemical Name or Material CCT 018159
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.