Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD 3254, Tocris Bioscience™

Catalog No. p-4783837 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective RXRα agonist.

Selective RXRα agonist; exhibits no activity at RARα, RARβ or RARγ receptors.

Chemical Identifiers

CAS 196961-43-0
Molecular Formula C24H28O3
Molecular Weight (g/mol) 364.485
InChI Key DYLLZSVPAUUSSB-VQHVLOKHSA-N
Synonym 3-4-hydroxy-3-3,5,5,8,8-pentamethyl-6,7-dihydronaphthalen, 3-4-hydroxy-3-3,5,5,8,8-pentamethyl-5,6,7,8-tetrahydronaphthalen-2-yl phenyl prop-2-enoic acid, 3-4-hydroxy-3-3,5,5,8,8-pentamethyl-6,7-dihydronaphthalen-2-yl phenyl prop-2-enoic acid
PubChem CID 44566110
IUPAC Name (E)-3-[4-hydroxy-3-(3,5,5,8,8-pentamethyl-6,7-dihydronaphthalen-2-yl)phenyl]prop-2-enoic acid
SMILES CC1=CC2=C(C=C1C3=C(C=CC(=C3)C=CC(=O)O)O)C(CCC2(C)C)(C)C

Specifications

Quantity 10 mg
Formula Weight 364.48
Percent Purity >97%
Chemical Name or Material CD 3254
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.