Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD 437, Tocris Bioscience™
GSA_VA

Catalog No. p-4783322 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

RARγ-selective agonist

Synthetic retinoid that is an RARγ-selective agonist. Displays RARγ-dependent and -independent effects on differentiation and apoptosis.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 125316-60-1
Molecular Formula C27H26O3
Molecular Weight (g/mol) 398.502
InChI Key LDGIHZJOIQSHPB-UHFFFAOYSA-N
Synonym ahpn, o-desmethyl adapalene, apoptosis activator vi, 6-3-1-adamantyl-4-hydroxyphenyl-2-naphthalene carboxylic acid, chembl1180, o-demethylated adapalene, 6-3-1-adamantyl-4-hydroxyphenyl-2-naphthalenecarboxylic acid, 2-naphthalenecarboxylicacid, 6-4-hydroxy-3-tricyclo 3.3.1.13,7 dec-1-ylphenyl, 6-4-hydroxy-3-tricyclo 3.3.1.1 3,7 dec-1-ylphenyl-2-naphthalenecarboxylic acid, 6-3-1-adamantyl-4-hydroxyphenyl naphthalene-2-carboxylic acid
PubChem CID 135411
IUPAC Name 6-[3-(1-adamantyl)-4-hydroxyphenyl]naphthalene-2-carboxylic acid
SMILES C1C2CC3CC1CC(C2)(C3)C4=C(C=CC(=C4)C5=CC6=C(C=C5)C=C(C=C6)C(=O)O)O

Specifications

Quantity 10 mg
Formula Weight 398.5
Percent Purity >98%
Chemical Name or Material CD 437
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.