Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD14 Mouse anti-Human, BUV395, Clone: M5E2, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 569102
Description
CD14 Monoclonal antibody specifically detects CD14 in Human samples. It is validated for Flow cytometrySpecifications
CD14 | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
929 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. | |
IgG2a κ |
Flow Cytometry | |
M5E2 | |
Brilliant Ultraviolet 395 | |
LPS receptor; LPS-R; Myeloid cell-specific leucine-rich glycoprotein | |
Human CD14 Protein | |
50 μg | |
Primary | |
Human | |
Liquid |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?