Learn More
CD1c Mouse anti-Human, BV650, Clone: F10/21A3, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 569617
Description
CD1c Monoclonal antibody specifically detects CD1c in Human samples. It is validated for Flow cytometrySpecifications
CD1c | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
911 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. | |
IgG1 κ |
Flow Cytometry | |
F10/21A3 | |
Brilliant Violet 650 | |
CD1; R7; M241; BDCA1 | |
GM-CSF- and IL-4-activated Human Monocytes | |
50 μg | |
Primary | |
Human | |
Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.