Learn More
CD20 Mouse anti-Human, RB780, Clone: 2H7, BD Horizon™
Mouse Monoclonal Antibody
$168.70 - $343.50
Specifications
Antigen | CD20 |
---|---|
Clone | 2H7 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | RB780 |
Description
CD20 Monoclonal antibody specifically detects CD20 in Human samples. It is validated for Flow cytometrySpecifications
CD20 | |
Flow Cytometry | |
RB780 | |
Mouse | |
Human | |
MS4A1; B1; Bp35; LEU-16; S7 | |
Human 6.16c1.3 B cell line | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. |
2H7 | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
931 | |
IgG2b κ | |
Affinity Chromatography |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.