Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD22 Mouse anti-Human, RB780, Clone: HIB22, BD Horizon™
Mouse Monoclonal Antibody
$205.00 - $307.50
Specifications
Antigen | CD22 |
---|---|
Clone | HIB22 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | RB780 |
Description
CD22 Monoclonal antibody specifically detects CD22 in Human samples. It is validated for Flow cytometrySpecifications
CD22 | |
Flow Cytometry | |
RB780 | |
Mouse | |
Human | |
BL-CAM; Siglec-2; Bgp135; Lyb8; LPAP | |
IgG1 κ | |
Affinity Chromatography |
HIB22 | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
933 | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. |
Spot an opportunity for improvement?
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.