Learn More
CD335 (NKp46) Rat anti-Mouse, BV650, Clone: 29A1.4, BD Horizon™
Rat Monoclonal Antibody
Supplier: BD Biosciences 569494
Description
CD335 (NKp46) Monoclonal antibody specifically detects CD335 (NKp46) in Mouse samples. It is validated for Flow cytometrySpecifications
CD335 (NKp46) | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
Affinity Chromatography | |
RUO | |
17086 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. | |
IgG2a κ |
Flow Cytometry | |
29A1.4 | |
Brilliant Violet 650 | |
Ncr1; NK-p46; NKp46; mNKp46; MAR1; mAR-1; mouse activating receptor 1; Ly94 | |
Mouse NKP46 Recombinant Protein | |
50 μg | |
Primary | |
Mouse | |
Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.