Learn More
STEMCELL Technologies EasySep™ Human CD4 Positive Selection Kit II, 1 x 10^9 cells
SDP
Supplier:  STEMCELL Technologies 17852
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
The EasySep™ Human CD4 Positive Selection Kit II is designed to isolate CD4+ cells from fresh or previously frozen peripheral blood mononuclear cells or washed leukapheresis samples by immunomagnetic positive selection. Desired cells are targeted with antibody complexes recognizing CD4 and magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.This product replaces the EasySep™ Human CD4 Positive Selection Kit (Catalog #18052) for even faster cell isolations.
Catalog No. NC1365354