Learn More
CD45 Mouse, anti-Human, RB780, Clone: HI30, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 568746
Description
The HI30 monoclonal antibody specifically binds to the 180, 190, 205, 220 kDa protein isoforms of CD45. CD45 is encoded by the PTPRC (Protein tyrosine phosphatase receptor type C) gene. CD45, also known as the leukocyte common antigen (LCA), is a member of the protein tyrosine phosphatase (PTP) family. It is present on all human leukocytes including lymphocytes, monocytes, granulocytes, eosinophils, and thymocytes. CD45 is absent from circulating erythrocytes, platelets, or mature erythroid cells of bone marrow and non-hemopoietic tissues.
Specifications
CD45 | |
HI30 | |
Aqueous buffered solution containing ≤0.09% sodium azide | |
Human Peripheral Blood Leucocytes | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
RB780 | |
Mouse | |
100 Tests | |
Secondary | |
Refrigerate (2°C to 8°C) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.