Learn More
CD45RA Mouse anti-Human, RB780, Clone: HI100, BD Horizon™
Mouse Monoclonal Antibody
$134.60 - $349.50
Specifications
Antigen | CD45RA |
---|---|
Clone | HI100 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | RB780 |
Description
CD45RA Monoclonal antibody specifically detects CD45RA in Human samples. It is validated for Flow cytometrySpecifications
CD45RA | |
Flow Cytometry | |
RB780 | |
Mouse | |
Human | |
CD45R; PTPRC; LCA; Leukocyte common antigen | |
IgG2b κ | |
Affinity Chromatography |
HI100 | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
5788 | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.