Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD62L Mouse anti-Human, BV786, Clone: SK11, BD Horizon™

Mouse Monoclonal Antibody

$224.00 - $442.00

Specifications

Antigen CD62L
Isotype Mouse BALB/c IgG2a, κ
Immunogen Human Peripheral Blood T Lymphocytes
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Conjugate Brilliant Violet 786
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB565312
View Documents
BD Biosciences
565312
25 Tests
Each for $224.00
Add to Cart
 
BDB565311
View Documents
BD Biosciences
565311
100 Tests
Each for $442.00
Add to Cart
 
Description

Description

CD62L Monoclonal antibody specifically detects CD62L in Baboon, Cynomolgus, Human, Rhesus Ma samples. It is validated for Flow cytometry.

CD62L Monoclonal antibody specifically detects CD62L in Baboon, Cynomolgus, Human, Rhesus Ma samples. It is validated for Flow cytometry.
Specifications

Specifications

CD62L
Human Peripheral Blood T Lymphocytes
Brilliant Violet 786
Cell Biology
Mouse
Purified
Monoclonal
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV786 under optimum conditions, and unconjugated antibody and free BD Horizon™ BV786 were removed.
Mouse BALB/c IgG2a, κ
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Primary
Flow Cytometry
RUO
SK11 (also known as Anti-Leu-8)
L-selectin; LECAM-1; SELL; LNHR; LSEL; LAM-1; LEU8; PLNHR; TQ-1; MEL-14
Vol. Per test: 5μL
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.