Learn More
ACROBiosystems ACROBiosystems Human CD7 Protein, His Tag
Supplier:  ACROBiosystems CD7H52H71MG
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Recombinant Protein;1MG;Human CD7, His Tag (CD7-H52H7) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Pro 180 (Accession # P09564-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 18.3 kDa. The protein migrates as 26-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6420