Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD73 Mouse anti-Human, RB780, Clone: AD2, BD Horizon™

Mouse Monoclonal Antibody

Supplier:  BD Biosciences 569218

 View more versions of this product

Catalog No. BDB569218


missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart

Description

Description

CD73 Monoclonal antibody specifically detects CD73 in Human samples. It is validated for Flow cytometry
Specifications

Specifications

CD73
Monoclonal
RB780
NT5E; 5' nucleotidase; 5'-NT; E5NT; Ecto-5'-nucleotidase; eN; eNT; NT; NT5
Pre-B leukemia cell line
25 Tests
Primary
Human
Liquid
Flow Cytometry
AD2
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse
Affinity Chromatography
RUO
4907
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed.
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.