Learn More
CD88 (C5a Receptor) Rat anti-Mouse, RB780, Clone: 20/70, BD OptiBuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 755755
Description
CD88 (C5a Receptor) Monoclonal antibody specifically detects CD88 (C5a Receptor) in Mouse samples. It is validated for Flow CytometrySpecifications
CD88 (C5a Receptor) | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
50 μg | |
Primary | |
Mouse | |
IgG2b κ |
Flow Cytometry | |
20/70 | |
RB780 | |
Cd88; C5ar1; C5a-R; C5aR; C5r1; C5a Ligand; C5a anaphylatoxin receptor | |
Mouse C5aR Transfected Cell Line | |
RUO | |
12273 | |
Store undiluted at 4°C and protected from prolonged exposure to light.Do not freeze.The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.