Learn More
CD88 Rat anti-Mouse, BV480, Clone: 20/70, BD OptiBuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 746588
Description
CD88 Monoclonal antibody specifically detects CD88 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD88 | |
20/70 | |
Brilliant Violet 480 | |
Rat | |
50 μg | |
Refrigerate (2 to 8°C) | |
Monoclonal |
Flow Cytometry | |
0.2 mg/mL | |
Purified | |
IgG2b, κ | |
RUO | |
Primary | |
Mouse |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.