Learn More
Abnova™ CD93 (Human) ELISA Kit
Shop All Abnova Corporation Products- CD93 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human CD93.
Specifications
Cross Reactivity
Human
Product Type
Kit
Description
CD93 molecule
Gene ID (Entrez)
22918
Storage Requirements
Store at 4°C for 6 months. Store at -20°C for 12 months. Avoid multiple freeze-thaw cycles.
Target
Human
Quantity
1 Kit
Gene Symbol
CD93
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.