Learn More
Abnova™ CDKN2D/CEN19p FISH Probe
Shop All Abnova Corporation ProductsSpecifications
Content And Storage
Store at 4°C in the dark.
Description
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
For Use With (Application)
FISH-Ce
Gene ID (Entrez)
1032
Includes
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
Quantity
100 μL
Format
Liquid
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.