Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDPPB, Tocris Bioscience™

Catalog No. p-4783838 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Positive allosteric modulator of mGlu5 receptors

Brain penetrant, selective positive allosteric modulator at the mGlu5 receptor (EC50 values are 10 and 20 nM for human and rat receptors respectively). Antipsychotic; reverses amphetamine-induced locomotor activity and amphetamine-induced deficits in prepulse inhibition in rats.

Chemical Identifiers

CAS 781652-57-1
Molecular Formula C23H16N4O
Molecular Weight (g/mol) 364.408
InChI Key BKUIZWILNWHFHD-UHFFFAOYSA-N
Synonym cdppb, 3-cyano-n-1,3-diphenyl-1h-pyrazol-5-yl benzamide, mglur5 ligand, cdppb, d0d4hg, d0m5hy, 3-cyano-n-2,5-di phenyl pyrazol-3-yl benzamide, 3-cyano-n-2,5-diphenyl-2h-pyrazol-3-yl-benzamide
PubChem CID 11245456
IUPAC Name 3-cyano-N-(2,5-diphenylpyrazol-3-yl)benzamide
SMILES C1=CC=C(C=C1)C2=NN(C(=C2)NC(=O)C3=CC=CC(=C3)C#N)C4=CC=CC=C4

Specifications

Quantity 10 mg
Formula Weight 364.4
Percent Purity >98%
Chemical Name or Material CDPPB
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.