Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Celastrol, Tocris Bioscience™

Catalog No. p-4783839 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Inhibits TNF-α-induced NF-κB activation

Antioxidant and anti-inflammatory agent. Potently inhibits lipid peroxidation in mitochondria and inhibits TNF-α-induced NFκB activation. Also shown to inhibit topoisomerase II activity in vitro (IC50 = 7.41 μM).

Chemical Identifiers

CAS 34157-83-0
Molecular Formula C29H38O4
Molecular Weight (g/mol) 450.62
MDL Number MFCD03424073
InChI Key KQJSQWZMSAGSHN-JJWQIEBTSA-N
Synonym celastrol, tripterine, tripterin, 2r,4as,6as,12br,14as,14br-10-hydroxy-2,4a,6a,9,12b,14a-hexamethyl-11-oxo-1,2,3,4,4a,5,6,6a,11,12b,13,14,14a,14b-tetradecahydropicene-2-carboxylic acid, triterpene, celastrol, celastrus scandens, unii-l8gg98663l, 3-hydroxy-24-nor-2-oxo-1 10 ,3,5,7-friedelatetraen-29-oic acid, 2r,4as,6ar,6as,14as,14br-10-hydroxy-2,4a,6a,6a,9,14a-hexamethyl-11-oxo-1,3,4,5,6,13,14,14b-octahydropicene-2-carboxylic acid, d:a-friedo-24-noroleana-1 10 ,3,5,7-tetraen-29-oic acid, 3-hydroxy-2-oxo-, 20alpha
PubChem CID 122724
ChEBI CHEBI:63959
IUPAC Name (2R,4aS,6aS,12bR,14aS,14bR)-10-hydroxy-2,4a,6a,9,12b,14a-hexamethyl-11-oxo-1,2,3,4,4a,5,6,6a,11,12b,13,14,14a,14b-tetradecahydropicene-2-carboxylic acid
SMILES CC1=C(O)C(=O)C=C2C1=CC=C1[C@@]2(C)CC[C@@]2(C)[C@@H]3C[C@@](C)(CC[C@]3(C)CC[C@]12C)C(O)=O

Specifications

Quantity 10 mg
Formula Weight 450.61
Percent Purity >99%
Chemical Name or Material Celastrol
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.