Learn More
Neogen Diagnostics Celecoxib Forensic ELISA Kit - 96 well
Supplier:  Neogen Diagnostics 180719
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Celecoxib ELISA is used for the quantitative analysis of celecoxib levels in urine, oral fluid, tissue culture supernatant, plasma or serum samples and is non-species specific. The kit was designed for screening purposes and is intended for forensic use only. Assay sample size: 20 µL.
Catalog No. NC2221775
missing translation for 'QuantityLeft'