Learn More
Bellco Glass, Inc. Cell Scraper Lifter 30mm 39cm
SDP
Supplier:  Bellco Glass, Inc. 229316000
Encompass_Preferred
Cell Scrapers are designed to gently harvest cells from tissue culture flasks dishes or bottles. Soft pliable blade won’t damage cells or scratch surfaces. Scraper blade is positioned parallel to the handle for windshield wiper like scraping. Lifter blade is positioned perpendicular to the handle for use with a raking motion. Pivoting blade Cell Scraper features free rotating blade that adjusts to the desired position providing access to all corners of the flask or tissue culture vessel.Specifications Handle Length 39cm Blade Length 30mm Blade Orientation Lifter Perpendicular Case Quantity 100
Catalog No. 50-232-9381
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.