Learn More
Bioworld Cell Scraper, Sterile, 25cm Handle 100/cs
Supplier: Bioworld 309000171
These cell scrapers allow for the gentle removal and harvesting of tissue cultures in all types of flasks, dishes and tubes. Handle Length: 25cm, Blade Length: 1.8cm, Polyethylene, Disposable, Gamma Radiation Sterilized, Non-Pyrogenic, Individually Wrapped. 100/case
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.