Learn More
Bellco Glass, Inc. Cell Strainer 40?m Blue
SDP
Supplier:  Bellco Glass, Inc. 229481000
Encompass_Preferred
CELLTREAT Cell Strainers can be used for many applications including isolating cells and removing clumps or debris. Polypropylene housing with nylon mesh filter eliminated the need to fabricate filters from gauze paper nylon mesh or other materials. Cell strainers are designed to universally fit most brands of 50mL conical tubes. The CELLTREAT exclusive pestle design improves manipulation of material in the strainer during use. Convex pestle head includes molded textured surface to better manipulate slippery material in the cell strainer.Specifications Extended lip on housing enables aseptic handling with forceps Elongated gripping tab on housing for easy handling Unique bulk packaging option available 5.5 inch pestle length for convenient hand manipulation Porosity Size 40um Color Blue Case Quantity 50
Catalog No. 50-233-0273
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.