Learn More
Alkali Scientific CellPro™ Aspirating Pipet, 2mL, Supplied without Graduations or Plugs, Individually Wrapped, Sterile, 200/cs
Supplier: Alkali Scientific SP202A
CellPro™ Aspirating Pipets are eBeam sterile, non-plugged, nongraduated, single-use pipets that can be used for a wide variety of vacuum-aspirating procedures, and are a safer alternative to glass pasteur pipets. Pipets are supplied in a thermoformed paper/plastic packaging wrap to protect against contamination. Nonpyrogenic.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.