Learn More
Alkali Scientific CellPro™ LB Lennox Agar, 10/Sleeve
Supplier: Alkali Scientific DC0046
CellPro™ LB Lennox Agar is a modification of Luria-Bertani (LB) agar, formulated with reduced sodium chloride content compared to standard LB agar. This medium is commonly used for molecular biology applications, including the cultivation and selection of recombinant bacterial strains. LB Lennox Agar provides a nutrient-rich environment for bacterial growth, supporting the propagation of transformed cells harboring plasmids with antibiotic resistance markers.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.