Learn More
Alkali Scientific CellPro™ LB (Lennox) Agar 150mm, 10/Sleeve
SDP
Supplier:  Alkali Scientific DC0142
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
CellPro™ LB Lennox Agar is a modification of Luria-Bertani (LB) agar, formulated with reduced sodium chloride content compared to standard LB agar. This medium is commonly used for molecular biology applications, including the cultivation and selection of recombinant bacterial strains. LB Lennox Agar provides a nutrient-rich environment for bacterial growth, supporting the propagation of transformed cells harboring plasmids with antibiotic resistance markers.
Catalog No. 50-238-9504