Learn More
Alkali Scientific CellPro™ LB (Lennox) + Gentamycin 50µg/mL 150mm
SDP
Supplier:  Alkali Scientific DC0161
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
CellPro™ LB (Lennox) + Gentamycin is a specialized culture medium containing gentamicin, an aminoglycoside antibiotic commonly used to select for bacterial strains carrying gentamicin resistance genes. This modified LB agar variant is suitable for the cultivation and maintenance of recombinant bacterial cells in molecular biology laboratories. By incorporating gentamicin into the LB agar, this medium enables the selective growth of transformed bacterial strains while inhibiting the growth of gentamicin-sensitive contaminants.
Catalog No. 50-238-8729
missing translation for 'QuantityLeft'