Learn More
Alkali Scientific CellPro™ SOC Media 500mL PC
SDP
Supplier:  Alkali Scientific DC0022
Encompass_Preferred
Alkali Scientific™ CellPro™ SOC Media is a specialized growth medium designed to support the transformation and propagation of Escherichia coli cells. Formulated with essential nutrients and salts, this media promotes rapid cell growth and high transformation efficiency, making it ideal for molecular cloning and recombinant DNA techniques. Through meticulous sterile filtration, CellPro™ SOC Media maintains purity and consistency, ensuring optimal performance in molecular biology laboratories.
Catalog No. 50-238-8728
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.