Learn More
Greiner Bio-One™ CellStar™ 96-Well, Cell Culture-Treated, V-Shaped-Bottom Microplate
Color: Clear; Sterility: Sterile; Lid: With Lid
$482.00 - $526.50
Specifications
No. of Wells 96
Color Clear
Sterility Sterile
View More Specs
Products 2
Catalog Number Mfr. No. dimPackaging Price Quantity & Availability  
Catalog Number Mfr. No. dimPackaging Price Quantity & Availability  
07-000-168
View Documents
Greiner Bio-One™
651180
Case of 100 for $526.50
missing translation for 'QuantityLeft'
Add to Cart
07-000-615
View Documents
Greiner Bio-One
651182
Case of 80 for $482.00
missing translation for 'QuantityLeft'
Add to Cart
Description
Description
  • Manufactured from clear polystyrene
  • Chimney well design reduces cross-contamination
Specifications
Specifications
96
Clear
Sterile
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.