Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium, 99.8% (metals basis), Thermo Scientific Chemicals

Catalog No. p-4824670
Click to view available options
Quantity:
1 g
5 g
50 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-46-2
Molecular Formula Cs
Molecular Weight (g/mol) 132.91
MDL Number MFCD00134037
InChI Key TVFDJXOCXUVLDH-UHFFFAOYSA-N
Synonym caesium, cesium-133, cesium, elemental, unii-1ksv9v4y4i, metals basis, 1ksv9v4y4i, cesio, zaesium, casium, zasium
PubChem CID 5354618
ChEBI CHEBI:30514
SMILES [Cs]

Specifications

Color Silver
Assay Percent Range 99.8% (Metals basis)
Quantity 1 g
IUPAC Name caesium
Chemical Name or Material Cesium
Hazard Category H260-H314-H361
Precautionary Statement P201-P202-P223-P231+P232-P260-P264b-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c
DOTInformation DOT Class: 4.3, Packaging Group: I
EINECSNumber 231-155-4
RTECSNumber FK9225000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.