Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cevimeline hydrochloride, Tocris Bioscience™

Catalog No. p-4816831 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective M1 agonist

Selective M1 receptor agonist. Induces atropine-sensitive contractions of isolated guinea pig ilea and trachea preparations (EC50 values are 3.5 and 3 μM respectively). Reverses AF64A-induced cognitive impairments in vivo.

Chemical Identifiers

CAS 107220-28-0
Molecular Formula C10H18ClNOS
Molecular Weight (g/mol) 235.77
InChI Key SURWTGAXEIEOGY-UHFFFAOYSA-N
Synonym hydrogen cevimeline chloride
PubChem CID 9837584
IUPAC Name 2-methylspiro[1,3-oxathiolane-5,3'-1-azabicyclo[2.2.2]octane];hydrochloride
SMILES CC1OC2(CN3CCC2CC3)CS1.Cl

Specifications

Quantity 10 mg
Formula Weight 235.77
Percent Purity >98%
Chemical Name or Material Cevimeline hydrochloride
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.