Learn More
Chondrex Inc CFA, 2 MG/ML X 5 ML
Supplier: Chondrex Inc 7009
Heat-killed M. tuberculosis H37 RA (non-viable) suspended in Incomplete Freund's Adjuvant at a concentration of 2 mg/ml. 5 mL.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.