Learn More
Thermo Scientific Dionex IonPac CS19 IC Columns | | 1 EA
Supplier:  Thermo Scientific 076029
Encompass
Separate polar amines such as ethanolamines using a high-capacity carboxylate-functionalized cation-exchange column with simple aqueous eluents. Specifically optimized for the separation of common cations, small polar amines, and moderately hydrophobic and polyvalent cations using simple aqueous eluents and elevated temperatures. Ideal for analysis of power plant waters treated with ammonium, morpholine, or ethanolamine, and can also be used to analyze chemical additives, chemical process solutions, scrubber solutions, personal care products, and food samples.
Catalog No. NC0640144
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.