Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CGP 52432, Tocris Bioscience™

Catalog No. p-4783333 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective GABAB antagonist

Potent, selective GABAB receptor antagonist (IC50 = 85 nM).

Chemical Identifiers

CAS 139667-74-6
Molecular Formula C15H24Cl2NO4P
Molecular Weight (g/mol) 384.234
InChI Key GJZVQXWEIYRHBE-UHFFFAOYSA-N
Synonym tocris-1246, biomol-nt_000239, 3-3,4-dichlorophenyl methyl amino propyl diethoxymethyl phosphinic acid, 3-3,4-dichlorphenyl methyl amino propyl diethoxymethyl phosphinic acid, 3-3,4-dichlorophenyl methylamino propyl-diethoxymethyl phosphinic acid, phosphinic acid, 3-3,4-dichlorophenyl methyl amino propyl diethoxymethyl
PubChem CID 132252
IUPAC Name 3-[(3,4-dichlorophenyl)methylamino]propyl-(diethoxymethyl)phosphinic acid
SMILES CCOC(OCC)P(=O)(CCCNCC1=CC(=C(C=C1)Cl)Cl)O

Specifications

Quantity 10 mg
Formula Weight 384.24
Percent Purity >99%
Chemical Name or Material CGP 52432
Recommended Storage Desiccate at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.