Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CGP 54626 hydrochloride, Tocris Bioscience™
GSA_VA

Catalog No. p-4783334 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective GABAB antagonist

A potent, selective GABAB receptor antagonist (IC50 = 4 nM).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 149184-21-4
Molecular Formula C18H29Cl3NO3P
Molecular Weight (g/mol) 444.758
InChI Key ZQCFHOVIXCJPLE-LINSIKMZSA-N
Synonym cgp 54626 hydrochloride, 3-n-1-3,4-dichlorophenyl ethylamino-2-hydroxypropyl cyclohexylmethyl phosphinic acid, s-r*,r*-3-1-3,4-dichlorophenyl ethyl amino-2-hydroxypropyl cyclohexylmethyl phosphinic acid, cyclohexylmethyl-2s-3-1s-1-3,4-dichlorophenyl ethyl amino-2-hydroxypropyl phosphinic acid hydrochloride
PubChem CID 197583
IUPAC Name cyclohexylmethyl-[(2S)-3-[[(1S)-1-(3,4-dichlorophenyl)ethyl]amino]-2-hydroxypropyl]phosphinic acid;hydrochloride
SMILES CC(C1=CC(=C(C=C1)Cl)Cl)NCC(CP(=O)(CC2CCCCC2)O)O.Cl

Specifications

Quantity 10 mg
Formula Weight 444.76
Percent Purity >96%
Chemical Name or Material CGP 54626 hydrochloride
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.