Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CGP 57380, Tocris Bioscience™

Catalog No. p-4783846 Shop All R&D Systems Products
Click to view available options
Quantity:
1 mg
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective inhibitor of Mnk1

Inhibitor of MAP-kinase interacting kinase-1 (Mnk1, MKNK1) (IC50 = 2.2 μM) that displays selectivity over p38, JNK1, ERK1, ERK2, PKC and c-src family kinases. Blocks phosphorylation of eIF4E in cellular assays (IC50 = 3 μM) and inhibits LPS-induced TNFα expression in macrophages.

Chemical Identifiers

CAS 522629-08-9
Molecular Formula C11H9FN6
Molecular Weight (g/mol) 244.233
InChI Key UQPMANVRZYYQMD-UHFFFAOYSA-N
Synonym mnk1 inhibitor, n3-4-fluorophenyl-1h-pyrazolo 3,4-d pyrimidine-3,4-diamine, 4-amino-5-4-fluoroanilino-pyrazolo 3,4-d pyrimidine, d0a7fq, cgp hplc, n-4-fluorophenyl-3h-pyrazolo 3,4-d pyrimidine-3,4-diamine, 3-n-4-fluorophenyl-1h-pyrazolo 3,4-d pyrimidine-3,4-diamine, 3-n-4-fluorophenyl-2h-pyrazolo 3,4-d pyrimidine-3,4-diamine, n3-4-fluorophenyl-1h-pyrazolo-3,4-d pyrimidine-3,4-diamine, n3-4-fluorophenyl-2h-pyrazolo 3,4-d pyrimidine-3,4-diamine
PubChem CID 11644425
IUPAC Name 3-N-(4-fluorophenyl)-2H-pyrazolo[3,4-d]pyrimidine-3,4-diamine
SMILES C1=CC(=CC=C1NC2=C3C(=NC=NC3=NN2)N)F

Specifications

Quantity 1 mg
Formula Weight 244.23
Percent Purity >98%
Chemical Name or Material CGP 57380
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.