Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CGP 7930, Tocris Bioscience™

Catalog No. p-4783337 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Positive allosteric modulator of GABAB receptors

Positive allosteric modulator of GABAB receptors. Increases the potency and efficacy of GABA at both native and recombinant GABAB receptors (EC50 values are 5.37 and 4.60μM respectively) and enhances the inhibitory effect of the agonist L-baclofen in cultured cortical neurons. Reduces operant self-administration of ethanol in alcohol-preferring rats following i.p. administration.

Chemical Identifiers

CAS 57717-80-3
Molecular Formula C19H32O2
Molecular Weight (g/mol) 292.463
InChI Key XLWJPQQFJNGUPA-UHFFFAOYSA-N
Synonym 2,6-di-tert-butyl-4-3-hydroxy-2,2-dimethylpropyl phenol, 2,6-ditert-butyl-4-3-hydroxy-2,2-dimethylpropyl phenol, tocris-1513, lopac-c-0862, cgp hplc , solid, 2,2-di-methyl-3-3,5-di-t-butyl-4-hydroxyphenyl propanol, 3-3,5-di-t-butyl-4-hydroxyphenyl-2,2-dimethylpropanol, 3,5-bis 1,1-dimethylethyl-4-hydroxy-?,?-dimethyl-benzenepropanol, 3-3',5'-di-tert-butyl-4'-hydroxy phenyl-2,2-dimethylpropanol, 3-3 inverted exclamation marka,5 inverted exclamation marka-di-tert-butyl-4 inverted exclamation marka-hydroxy phenyl-2,2-dimethylpropanol
PubChem CID 5024764
IUPAC Name 2,6-ditert-butyl-4-(3-hydroxy-2,2-dimethylpropyl)phenol
SMILES CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)CC(C)(C)CO

Specifications

Quantity 10 mg
Formula Weight 292.46
Percent Purity >98%
Chemical Name or Material CGP 7930
Recommended Storage Desiccate at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.