Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CGS 12066B dimaleate, Tocris Bioscience™

Catalog No. p-4783338 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

5-HT1B agonist

5-HT1B full agonist, 10-fold selective over 5-HT1A and 1000-fold selective over 5-HT2C receptors. Centrally active following systemic administration.

Chemical Identifiers

CAS 109028-10-6
Molecular Formula C25H25F3N4O8
Molecular Weight (g/mol) 566.49
InChI Key HTEVMLYDEWVIQE-LVEZLNDCSA-N
Synonym cgs 12066b dimaleate, 1-methyl-4-7-trifluoromethyl pyrrolo 1,2-a quinoxalin-4-yl piperazine; bis butenedioic acid
PubChem CID 71299720
IUPAC Name (E)-but-2-enedioic acid;4-(4-methylpiperazin-1-yl)-7-(trifluoromethyl)pyrrolo[1,2-a]quinoxaline
SMILES CN1CCN(CC1)C2=NC3=C(C=CC(=C3)C(F)(F)F)N4C2=CC=C4.C(=CC(=O)O)C(=O)O.C(=CC(=O)O)C(=O)O

Specifications

Quantity 10 mg
Formula Weight 566.49
Percent Purity >98%
Chemical Name or Material CGS 12066B dimaleate
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.