Learn More
TARGETMOL CHEMICALS INC CGS 15943 10MG
Supplier:  TARGETMOL CHEMICALS INC T14944
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CGS 15943 is an orally bioavailable non-xanthine antagonist of Adenosine Receptor. In transfected CHO cells its Ki for human A1 A2A A2B and A3 Adenosine Receptors are 3.5 4.2 16 and 50 nM respectively. purity: 99%
Catalog No. 50-257-7369
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.