Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CGS 15943, Tocris Bioscience™

Catalog No. p-4783847 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent adenosine receptor antagonist

CGS 15943 is a potent adenosine receptor antagonist (Ki values are 3.5, 4.2, 16 and 51 nM for human A1, A2A, A2B and A3 receptors respectively). Inhibits the catalytic subunit of the class IB PI3K isoform p110γ (IC50 = 1.1 μM). Blocks HCC and HDAC cell proliferation in vitro via inhibition of the PI3K/Akt pathway; reduces proliferation of ER+ breast cancer cells. Also acts as a BMP-4 mimetic to stimulate osteogenic differentiation. Orally active in vivo.

Chemical Identifiers

CAS 104615-18-1
Molecular Formula C13H8ClN5O
Molecular Weight (g/mol) 285.69
MDL Number MFCD01529897
InChI Key MSJODEOZODDVGW-UHFFFAOYSA-N
Synonym 9-chloro-2-2-furyl-1,2,4 triazolo 1,5-c quinazolin-5-imine, 9-chloro-2-furan-2-yl-1,2,4 triazolo 1,5-c quinazolin-5-amine, chembl16687, 9-chloro-2-2-furyl 1,2,4 triazolo 1,5-c quinazolin-5-amine, 1,2,4 triazolo 1,5-c quinazolin-5-amine,9-chloro-2-2-furanyl, 9-chloro-2-2-furanyl-1,2,4 triazolo 1,5-c quinazolin-5-amine, 1,2,4 triazolo 1,5-c quinazolin-5-amine, 9-chloro-2-2-furanyl, acmc-20ddlj
PubChem CID 2690
IUPAC Name 9-chloro-2-(furan-2-yl)-[1,2,4]triazolo[1,5-c]quinazolin-5-amine
SMILES NC1=NC2=CC=C(Cl)C=C2C2=NC(=NN12)C1=CC=CO1

Specifications

Quantity 10 mg
Formula Weight 285.69
Percent Purity >99%
Chemical Name or Material CGS 15943
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.