Learn More
TARGETMOL CHEMICALS INC CGS 21680 HYDROCHLORIDE 10MG
Supplier:  TARGETMOL CHEMICALS INC T6441
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CGS 21680 Hydrochloride (CGS 21680 HCl)(IC50=22 nM) an adenosine receptor agonist exhibits 140-fold potency in A2 receptor over A1 receptor. purity: 98%
Catalog No. 50-257-6517
missing translation for 'QuantityLeft'
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.